missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BTN1A1/Butyrophilin Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB84671-100
This item is not returnable.
View return policy
Description
BTN1A1/Butyrophilin Monoclonal specifically detects BTN1A1/Butyrophilin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.
Specifications
| BTN1A1/Butyrophilin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| BTBTNbK14H9.2 (butyrophilin, subfamily 1, member A1), BTN, BTN1A1, butyrophilin subfamily 1 member A1, butyrophilin, subfamily 1, member A1 | |
| Mouse myeloma cell line NS0-derived recombinant human BTN1A1/Butyrophilin Ala27-Arg242 Accession # Q13410 | |
| 100 μg | |
| Primary | |
| Detects human BTN1A1/Butyrophilin in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry | |
| 2151B | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 0.3-25 ug/mL | |
| Q13410 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 696 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction