missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BTN1A1/Butyrophilin Antibody, R&D Systems™

Rabbit Monoclonal Antibody
209.00€ - 500.00€
Specifications
| Antigen | BTN1A1/Butyrophilin |
|---|---|
| Clone | 2151B |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 0.3-25 ug/mL |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18704088
![]() |
R&D Systems
MAB84671-SP |
25 μg |
209.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18758169
![]() |
R&D Systems
MAB84671-100 |
100 μg |
500.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BTN1A1/Butyrophilin Monoclonal specifically detects BTN1A1/Butyrophilin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
| BTN1A1/Butyrophilin | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 0.3-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| BTBTNbK14H9.2 (butyrophilin, subfamily 1, member A1), BTN, BTN1A1, butyrophilin subfamily 1 member A1, butyrophilin, subfamily 1, member A1 | |
| Mouse myeloma cell line NS0-derived recombinant human BTN1A1/Butyrophilin Ala27-Arg242 Accession # Q13410 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human BTN1A1/Butyrophilin in direct ELISAs. |
| 2151B | |
| Flow Cytometry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q13410 | |
| 696 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title