missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR3 PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 9 publications
Brand: R&D Systems FAB155P-025
This item is not returnable.
View return policy
Description
CCR3 Monoclonal specifically detects CCR3 in Human samples. It is validated for Flow Cytometry.
Specifications
| CCR3 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR3, CCR-3, CD193, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1232 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 61828 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P51677 | |
| CCR3 | |
| Y3 rat myeloid cell line transfected with human CCR3 Accession # P51677 | |
| 25 Tests | |
| Primary | |
| Detects human CCR3 transfectants in flow cytometry. In flow cytometry, no cross-reactivity with CCR1, CCR2, CCR5, and CCR6 transfectants or the parental cell line is observed. Detects CCR3 on peripheral blood eosinophils. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction