missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR3 PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 9 publications
217.00€ - 492.00€
Specifications
| Antigen | CCR3 |
|---|---|
| Clone | 61828 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18758128
![]() |
R&D Systems
FAB155P-025 |
25 Tests |
217.00€
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18715776
![]() |
R&D Systems
FAB155P-100 |
100 Tests |
492.00€
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR3 Monoclonal specifically detects CCR3 in Human samples. It is validated for Flow Cytometry.Specifications
| CCR3 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR3, CCR-3, CD193, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841 | |
| CCR3 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human CCR3 transfectants in flow cytometry. In flow cytometry, no cross-reactivity with CCR1, CCR2, CCR5, and CCR6 transfectants or the parental cell line is observed. Detects CCR3 on peripheral blood eosinophils. |
| 61828 | |
| Flow Cytometry | |
| R-PE | |
| Rat | |
| Human | |
| P51677 | |
| 1232 | |
| Y3 rat myeloid cell line transfected with human CCR3 Accession # P51677 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title