missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Component C1rLP Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3676-SP
This item is not returnable.
View return policy
Description
Complement Component C1rLP Polyclonal specifically detects Complement Component C1rLP in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Complement Component C1rLP | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C1RL, C1RL1, C1RL1complement C1r subcomponent-like protein, C1r-like protein, C1r-like serine protease analog protein, C1r-LPcomplement C1r-like proteinase, C1RLPEC 3.4.21.-, CLSPa, complement component 1, r subcomponent-like, EC 3.4.21, EC 3.4.21.41 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 51279 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| AAI56781 | |
| C1RL | |
| Mouse myeloma cell line NS0-derived recombinant human Complement Component C1rLP Ser41-Asn487 Accession # AAI56781 | |
| 25 μg | |
| Primary | |
| Detects human Complement Component C1rLP in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant mouse C1rLP is observed and less than 5% cross-reactivity with recombinant human (rh) C1R, rhC1S, and rhMASP-3 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction