missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Component C1rLP Antibody, R&D Systems™

Goat Polyclonal Antibody
196.00€ - 472.00€
Specifications
| Antigen | Complement Component C1rLP |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18776197
![]() |
R&D Systems
AF3676-SP |
25 μg |
196.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18058471
![]() |
R&D Systems
AF3676 |
100 μg |
472.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Complement Component C1rLP Polyclonal specifically detects Complement Component C1rLP in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Complement Component C1rLP | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| C1RL, C1RL1, C1RL1complement C1r subcomponent-like protein, C1r-like protein, C1r-like serine protease analog protein, C1r-LPcomplement C1r-like proteinase, C1RLPEC 3.4.21.-, CLSPa, complement component 1, r subcomponent-like, EC 3.4.21, EC 3.4.21.41 | |
| C1RL | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| AAI56781 | |
| 51279 | |
| Mouse myeloma cell line NS0-derived recombinant human Complement Component C1rLP Ser41-Asn487 Accession # AAI56781 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Complement Component C1rLP in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant mouse C1rLP is observed and less than 5% cross-reactivity with recombinant human (rh) C1R, rhC1S, and rhMASP-3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title