missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cystatin D Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1202
This item is not returnable.
View return policy
Description
Cystatin D/Cst5 Polyclonal specifically detects Cystatin D/Cst5 in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| Cystatin D/Cst5 | |
| Polyclonal | |
| Unconjugated | |
| CAA49838 | |
| CST5 | |
| Mouse myeloma cell line NS0-derived recombinant human Cystatin D Thr29-Val142 Accession # CAA49838 | |
| 100 μg | |
| Primary | |
| Detects human Cystatin D in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human (rh) Cystatin C, rhCystatin SN, and rhCystatin S is observed and less than 1% cross-reactivity with rhCystatin E/M and rhFetuin A is observed. | |
| Human | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 13 ug/mL | |
| CST5, CSTD, cystatin 5, cystatin D, Cystatin-5, cystatin-D, cysteine-proteinase inhibitor, MGC71922 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1473 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction