missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cystatin D Antibody, R&D Systems™

Goat Polyclonal Antibody
209.00€ - 569.00€
Specifications
| Antigen | Cystatin D/Cst5 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Neutralization 13 ug/mL |
| Applications | Western Blot, Neutralization |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18775807
![]() |
R&D Systems
AF1202-SP |
25 μg |
209.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18048911
|
R&D Systems
AF1202 |
100 μg |
569.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cystatin D/Cst5 Polyclonal specifically detects Cystatin D/Cst5 in Human samples. It is validated for Western Blot, Neutralization.Specifications
| Cystatin D/Cst5 | |
| Western Blot 0.1 ug/mL, Neutralization 13 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| CST5, CSTD, cystatin 5, cystatin D, Cystatin-5, cystatin-D, cysteine-proteinase inhibitor, MGC71922 | |
| CST5 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Neutralization | |
| Unconjugated | |
| RUO | |
| CAA49838 | |
| 1473 | |
| Mouse myeloma cell line NS0-derived recombinant human Cystatin D Thr29-Val142 Accession # CAA49838 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Cystatin D in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human (rh) Cystatin C, rhCystatin SN, and rhCystatin S is observed and less than 1% cross-reactivity with rhCystatin E/M and rhFetuin A is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title