missing translation for 'onlineSavingsMsg'
Learn More

Human Exosome Component 9 Antibody, R&D Systems™

Mouse Monoclonal Antibody

Brand:  R&D Systems MAB11616-SP

 View more versions of this product

Product Code. 30279915

  • 234.00€ / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Exosome Component 9 Monoclonal antibody specifically detects Exosome Component 9 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Exosome Component 9
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Autoantigen PM/Scl 1, EC 3.1.13, exosome complex component RRP45, exosome complex exonuclease RRP45, P5, p6, P75 polymyositis-scleroderma overlap syndrome associated autoantigen, PCH1D, PM/Scl-75, PMSCL autoantigen, 75kD, PMSCL1, Polymyositis/scleroderma autoantigen 1, polymyositis/scleroderma autoantigen 1 (75kD), polymyositis/scleroderma autoantigen 1, 75kDa, RRP45, Rrp45p
Synthetic Peptide, Accession # Q06265
25 μg
Primary
Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution.
IgG1
Immunohistochemistry, Immunocytochemistry
1090209
Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL
Q06265
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
5393
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.