missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF basic/FGF2/bFGF Antibody, R&D Systems™

Description
FGF basic/FGF2/bFGF Polyclonal specifically detects FGF basic/FGF2/bFGF in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
Specifications
Specifications
| Antigen | FGF basic/FGF2 |
| Applications | Western Blot, Immunohistochemistry, Neutralization |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.08-0.4 ug/mL |
| Gene Alias | Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin |
| Gene Symbols | FGF2 |
| Host Species | Goat |
| Immunogen | Bovine brain-derived FGF basic/FGF2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?