missing translation for 'onlineSavingsMsg'
Learn More

Human Fgr Antibody, R&D Systems™

Product Code. 18180851
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18180851 100 μg 100µg
18705209 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18180851 Supplier R&D Systems Supplier No. MAB32071

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fgr Monoclonal specifically detects Fgr in Human samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fgr
Applications Immunocytochemistry
Classification Monoclonal
Clone 349331
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P09769
Gene Alias c-fgr, c-src2, c-src-2 proto-oncogene, EC 2.7.10, EC 2.7.10.2, FLJ43153, Gardner-Rasheed feline sarcoma viral (v-fgr) oncogene homolog, MGC75096, p55c-fgr, p55-c-fgr protein, p55-Fgr, p58c-fgr, Proto-oncogene c-Fgr, proto-oncogene tyrosine-protein kinase FGR, Src2, SRC2c-fgr protooncogene, tyrosine-protein kinase Fgr
Gene Symbols FGR
Host Species Mouse
Immunogen E. coli-derived recombinant human Fgr Met1-Phe82 Accession # P09769
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2268
Test Specificity Detects human Fgr in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.