Learn More
Human IFN-gamma R1/CD119 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 4 publications
Brand:  R&D Systems MAB6732-500
 View more versions of this product
Product Code. 18778438
  • 2070.00€ / 500µg
Description
IFN-gamma R1/CD119 Monoclonal specifically detects IFN-gamma R1/CD119 in Human samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFN-gamma R1/CD119
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
IFNGR1
Human placenta-derived IFN-gamma R1/CD119
500 μg
Primary
Detects human IFN-gamma R1/CD119 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse IFN-gamma R1, recombinant human (rh) IFN-gamma R2, or rhIL-10 R beta is observed.
Human
Supernatant
Neutralization
GIR208
Neutralization 0.03-0.15 ug/mL
AVP, type 2, CD119, CD119 antigen, FLJ45734, IFN-gamma receptor 1, IFNgammaR1, IFN-gamma-R1, IFNGR, IFN-gR1, IFNGR1, immune interferon receptor 1, interferon gamma receptor 1, interferon-gamma receptor alpha chain, type 2
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3459
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.