missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-gamma R1/CD119 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
211.00€ - 2070.00€
Specifications
| Antigen | IFN-gamma R1/CD119 |
|---|---|
| Clone | GIR208 |
| Dilution | Neutralization 0.03-0.15 ug/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18762848
![]() |
R&D Systems
MAB6732-SP |
25 μg |
211.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18715158
![]() |
R&D Systems
MAB6732-100 |
100 μg |
532.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18778438
![]() |
R&D Systems
MAB6732-500 |
500 μg |
2070.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-gamma R1/CD119 Monoclonal specifically detects IFN-gamma R1/CD119 in Human samples. It is validated for Neutralization.Specifications
| IFN-gamma R1/CD119 | |
| Neutralization 0.03-0.15 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| AVP, type 2, CD119, CD119 antigen, FLJ45734, IFN-gamma receptor 1, IFNgammaR1, IFN-gamma-R1, IFNGR, IFN-gR1, IFNGR1, immune interferon receptor 1, interferon gamma receptor 1, interferon-gamma receptor alpha chain, type 2 | |
| IFNGR1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| GIR208 | |
| Neutralization | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 3459 | |
| Human placenta-derived IFN-gamma R1/CD119 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IFN-gamma R1/CD119 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse IFN-gamma R1, recombinant human (rh) IFN-gamma R2, or rhIL-10 R beta is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title