missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-12 p70 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB611R-100
This item is not returnable.
View return policy
Description
IL-12 p70/IL-12A Monoclonal specifically detects IL-12 p70/IL-12A in Human samples. It is validated for Immunocytochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| IL-12 p70/IL-12A | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CLMF p35, CLMF1, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL12, IL12 p70, IL-12 subunit p35, IL-12, subunit p35, IL12A, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-12 heterodimer Arg23-Ser219 of p35, Ile23-Ser328 of p40 Accession # P29459(p35) & P29460 (p40) | |
| 100 μg | |
| Primary | |
| Detects human IL-12 p70 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human (rh) IL-12 p40, rhIL-12 p35, rhIL-23 or recombinant mouse IL-12 is observed. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Immunocytochemistry, ELISA | |
| 24945R | |
| Immunocytochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P29459(p35) & P29460 (p40) | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3592 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?Share a Content Correction