missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-12 p70 Antibody, R&D Systems™

Mouse Monoclonal Antibody
249.00€ - 1775.00€
Specifications
| Antigen | IL-12 p70/IL-12A |
|---|---|
| Clone | 24945R |
| Dilution | Immunocytochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Immunocytochemistry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18764628
![]() |
R&D Systems
MAB611R-SP |
25 μg |
249.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18725408
![]() |
R&D Systems
MAB611R-100 |
100 μg |
451.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18199860
|
R&D Systems
MAB611R-500 |
500 ug |
1775.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18109460
|
R&D Systems
MAB611R-01M |
1 mg |
1595.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-12 p70/IL-12A Monoclonal specifically detects IL-12 p70/IL-12A in Human samples. It is validated for Immunocytochemistry, ELISA Capture (Matched Antibody Pair).Specifications
| IL-12 p70/IL-12A | |
| Immunocytochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| P29459(p35) & P29460 (p40) | |
| 3592 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 24945R | |
| Immunocytochemistry, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| CLMF p35, CLMF1, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL12, IL12 p70, IL-12 subunit p35, IL-12, subunit p35, IL12A, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Detects human IL-12 p70 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human (rh) IL-12 p40, rhIL-12 p35, rhIL-23 or recombinant mouse IL-12 is observed. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title