missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse FABP2/I-FABP Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18712067
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18712067 25 μg 25µg
18798859 100 μg 100µg
18750407 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18712067 Supplier R&D Systems Supplier No. MAB30781SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FABP2/I-FABP Monoclonal specifically detects FABP2/I-FABP in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FABP2/I-FABP
Applications Western Blot, ELISA
Classification Monoclonal
Clone 323730
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P12104
Gene Alias FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, IFABP, I-FABP, I-FABPMGC133132, Intestinal FABP, Intestinal-type fatty acid-binding protein
Gene Symbols FABP2
Host Species Mouse
Immunogen E. coli-derived recombinant human FABP2/I-FABP Ala2-Asp132 Accession # P12104
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2169
Test Specificity Detects human FABP2/I-FABP in ELISAs. Detects human and mouse FABP2/I-FABP in Western blots. In sandwich immunoassays, no cross-reactivity or interference with recombinant human FABP1, 3, 5, 6, 7, 8, 9, recombinant mouse FABP4, 9, or recombinant rat FABP2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.