missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Neurexin 3/NRXN3 Biotinylated Antibody, R&D Systems™

Description
Neurexin 3/NRXN3 Polyclonal specifically detects Neurexin 3/NRXN3 in Human, Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | Neurexin 3/NRXN3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Biotin |
| Dilution | Western Blot |
| Gene Alias | C14orf60, chromosome 14 open reading frame 60, KIAA0743MGC176711, neurexin 3, neurexin III, Neurexin III-alpha, Neurexin III-beta, neurexin-3-beta, NRXN3 |
| Gene Symbols | NRXN3 |
| Host Species | Sheep |
| Immunogen | Mouse myeloma cell line NS0-derived recombinant human Neurexin 3/NRXN3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?