missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Notch-2 Intracellular Domain Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3735-SP
This item is not returnable.
View return policy
Description
Notch-2 Monoclonal specifically detects Notch-2 in Human samples. It is validated for Western Blot.
Specifications
| Notch-2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q04721.3 | |
| NOTCH2 | |
| E. coli-derived recombinant human Notch-2 Asp2063-Pro2413 Accession # Q04721.3 | |
| 25 μg | |
| Primary | |
| Detects human Notch-2 intracellular domain (ICD) in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) Notch-1 ICD, recombinant rat (rr) Notch-1 ECD, rrNotch-2 ECD, rhNotch-3 ECD, or rhNotch-4 ICD. | |
| Human | |
| Purified |
| Western Blot | |
| 411801 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2Notch homolog 2 (Drosophila), Notch homolog 2, Notch2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4853 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction