missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Notch-2 Intracellular Domain Antibody, R&D Systems™

Rat Monoclonal Antibody
201.00€ - 396.00€
Specifications
| Antigen | Notch-2 |
|---|---|
| Clone | 411801 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18777729
![]() |
R&D Systems
MAB3735-SP |
25 μg |
201.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18121561
|
R&D Systems
MAB3735 |
100 μg |
396.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Notch-2 Monoclonal specifically detects Notch-2 in Human samples. It is validated for Western Blot.Specifications
| Notch-2 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Human | |
| Q04721.3 | |
| 4853 | |
| E. coli-derived recombinant human Notch-2 Asp2063-Pro2413 Accession # Q04721.3 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Notch-2 intracellular domain (ICD) in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) Notch-1 ICD, recombinant rat (rr) Notch-1 ECD, rrNotch-2 ECD, rhNotch-3 ECD, or rhNotch-4 ICD. |
| 411801 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2Notch homolog 2 (Drosophila), Notch homolog 2, Notch2 | |
| NOTCH2 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title