missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PGLYRP4/PGRP-I beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3018-SP
This item is not returnable.
View return policy
Description
PGLYRP4/PGRP-I beta Monoclonal specifically detects PGLYRP4/PGRP-I beta in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| PGLYRP4/PGRP-I beta | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q96LB8 | |
| PGLYRP4 | |
| Mouse myeloma cell line NS0-derived recombinant human PGLYRP4/PGRP-I beta Asp18-His373 Accession # Q96LB8 | |
| 25 μg | |
| Primary | |
| Detects human PGLYRP4/PGRP-I beta in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse PGRP-I beta or recombinant human PGRP-S is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 474034 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| peptidoglycan recognition protein 4, peptidoglycan recognition protein I beta, Pglyrp-1 beta, PGRPI beta, PGRP-I beta, PGRP-I-beta | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 57115 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction