missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PGLYRP4/PGRP-I beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
176.00€ - 377.00€
Specifications
| Antigen | PGLYRP4/PGRP-I beta |
|---|---|
| Clone | 474034 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18797669
![]() |
R&D Systems
MAB3018-SP |
25 μg |
176.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18190691
![]() |
R&D Systems
MAB3018 |
100 μg |
377.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PGLYRP4/PGRP-I beta Monoclonal specifically detects PGLYRP4/PGRP-I beta in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| PGLYRP4/PGRP-I beta | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q96LB8 | |
| 57115 | |
| Mouse myeloma cell line NS0-derived recombinant human PGLYRP4/PGRP-I beta Asp18-His373 Accession # Q96LB8 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human PGLYRP4/PGRP-I beta in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse PGRP-I beta or recombinant human PGRP-S is observed. |
| 474034 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| peptidoglycan recognition protein 4, peptidoglycan recognition protein I beta, Pglyrp-1 beta, PGRPI beta, PGRP-I beta, PGRP-I-beta | |
| PGLYRP4 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title