missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRA-1-81 PE-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems FAB8495P-100
This item is not returnable.
View return policy
Description
TRA-1-81 Monoclonal specifically detects TRA-1-81 in Human samples. It is validated for Flow Cytometry.
Specifications
| TRA-1-81 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| PODXL | |
| Human embryonal carcinoma cell line 2102Ep | |
| 100 Tests | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| TRA-1-81 | |
| Flow Cytometry 10 uL/10^6 cells | |
| gp200, MGC138240, PC, PCLP, PODXL, TRA181 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects human TRA-1-81 in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction