missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRA-1-81 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
277.00€ - 496.00€
Specifications
| Antigen | TRA-1-81 |
|---|---|
| Clone | TRA-1-81 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18134801
|
R&D Systems
FAB8495P-100 |
100 Tests |
496.00€
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18797889
![]() |
R&D Systems
FAB8495P-025 |
25 Tests |
277.00€
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRA-1-81 Monoclonal specifically detects TRA-1-81 in Human samples. It is validated for Flow Cytometry.Specifications
| TRA-1-81 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| PODXL | |
| IgM | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human TRA-1-81 in flow cytometry. |
| TRA-1-81 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| Human | |
| gp200, MGC138240, PC, PCLP, PODXL, TRA181 | |
| Human embryonal carcinoma cell line 2102Ep | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title