missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VE-Statin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF3638-SP
This item is not returnable.
View return policy
Description
VE-Statin/EGFL7 Polyclonal specifically detects VE-Statin/EGFL7 in Human samples. It is validated for Western Blot.
Specifications
| VE-Statin/EGFL7 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EGFL7, EGF-like protein 7, EGF-like-domain, multiple 7, epidermal growth factor-like protein 7, MEGF7, MGC111117, Multiple EGF-like domains protein 7, Multiple epidermal growth factor-like domains protein 7, NEU1 protein, NOTCH4-like protein, RP11-251M1.2, Vascular endothelial statin, VEStatin, VE-STATIN, Zneu1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 51162 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| BAD01469 | |
| EGFL7 | |
| E. coli-derived recombinant human VE-Statin Glu21-Ser273 Accession # BAD01469 | |
| 25 μg | |
| Primary | |
| Detects human VE-Statin in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse VE-Statin is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction