missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VE-Statin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
221.00€ - 612.00€
Specifications
| Antigen | VE-Statin/EGFL7 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18788647
![]() |
R&D Systems
AF3638-SP |
25 μg |
221.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18092322
|
R&D Systems
AF3638 |
100 μg |
612.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VE-Statin/EGFL7 Polyclonal specifically detects VE-Statin/EGFL7 in Human samples. It is validated for Western Blot.Specifications
| VE-Statin/EGFL7 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| EGFL7, EGF-like protein 7, EGF-like-domain, multiple 7, epidermal growth factor-like protein 7, MEGF7, MGC111117, Multiple EGF-like domains protein 7, Multiple epidermal growth factor-like domains protein 7, NEU1 protein, NOTCH4-like protein, RP11-251M1.2, Vascular endothelial statin, VEStatin, VE-STATIN, Zneu1 | |
| EGFL7 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| BAD01469 | |
| 51162 | |
| E. coli-derived recombinant human VE-Statin Glu21-Ser273 Accession # BAD01469 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human VE-Statin in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse VE-Statin is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title