missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Humanin Polyclonal Antibody

Description
Humanin Polyclonal Antibody for Western Blot
Humanin is a novel 24 A. A polypeptide identified in Alzheimer's disease-affected brain. It is 99% homologous to mitochondrial 16s ribosomal RNA and functions as a novel neuroprotective and cytoprotective factor. It specifically inhibits neuronal cell death induced by beta-amyloids and Alzheimer's disease causing FAD genes by competitively inhibiting access of FPRL1 receptor to beta-amyloids. It acts as an anti-apoptotic factor by binding to Bcl-2/Bax family of pro-apoptotic factors including Bax, Bak, Bid and BimEL and inhibits their pro-apoptotic activity. It activates JAK/STAT signaling pathway and is regulated by TRIM11. It interacts with IGFBP-3 and specifically blocks IGFBP-3 induced cell death in glioblastoma cells. Humanin is also expressed in colon and testis.
Specifications
Specifications
| Antigen | Humanin |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | MT-RNR2 |
| Gene Accession No. | Q8IVG9 |
| Gene Alias | BCL2L4; HN1; Humanin-like 1; humanin-like protein 1; mitochondrially encoded 16S RNA; MTRNR2; MT-RNR2; MTRNR2L1; MT-RNR2-like 1; MTRNR2-like 1; MT-RNR2-like protein 1; RNR2 |
| Gene Symbols | MT-RNR2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?