missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylapatite, MP Biomedicals™

Product Code. 11406622 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
Unit Size:
100g
10g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11406622 10 g 10g
11416622 25 g 25g
11426622 50 g 50g
11436622 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11406622 Supplier MP Biomedicals Supplier No. 0215016210
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1306-06-5
Molecular Formula Ca5HO13P3
Molecular Weight (g/mol) 502.31
MDL Number MFCD00010904
InChI Key XYJRXVWERLGGKC-UHFFFAOYSA-D
Synonym Calcium phosphate hydroxide, Durapatite
IUPAC Name pentacalcium hydroxide triphosphate
SMILES [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 10 g
Formula Weight 502.31
Chemical Name or Material Hydroxylapatite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.