missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropanedial, TRC

Product Code. 30133358
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133358 25 mg 25mg
30132414 10 mg 10mg
30008102 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133358 Supplier Toronto Research Chemicals Supplier No. TRCH95247525MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-15-4
Molecular Formula C3 H4 O3
Molecular Weight (g/mol) 88.06
IUPAC Name 2-hydroxypropanedial
SMILES OC(C=O)C=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C3H4O3/c4-1-3(6)2-5/h1-3,6H
Formula Weight 88.016
Chemical Name or Material Hydroxypropanedial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.