missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFI27 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16619894
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16619894 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16619894 Supplier Invitrogen™ Supplier No. PA5103440

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IFI27.

Promotes cell death. Mediates IFN-induced apoptosis characterized by a rapid and robust release of cytochrome C from the mitochondria and activation of BAX and caspases 2, 3, 6, 8 and 9.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFI27
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IFI27
Gene Accession No. P40305
Gene Alias 2310061N23Rik; FAM14D; IFI27; interferon alpha inducible protein 27; interferon alpha-induced 11.5 kDa protein; Interferon alpha-inducible protein 27, mitochondrial; interferon, alpha-inducible protein 27; interferon-stimulated gene 12a protein; ISG12; ISG12(a); ISG12A; p27
Gene Symbols IFI27
Host Species Rabbit
Immunogen A synthesized peptide derived from human IFI27(Accession P40305), corresponding to amino acid residues M1-I33.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3429
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.