missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFIT3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12668-100ul
This item is not returnable.
View return policy
Description
IFIT3 Polyclonal antibody specifically detects IFIT3 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| IFIT3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 5-10uL/mg of uLysate | |
| CIG-49, GARG-49, IFI60Retinoic acid-induced gene G protein, IFIT-3, IFIT-4, IFIT4IFI-60K, interferon-induced protein with tetratricopeptide repeats 3, Interferon-induced protein with tetratricopeptide repeats 4Interferon-induced 60 kDa protein, IRG2, ISG60, ISG-60, RIG-GCIG49 | |
| Produced in rabbits immunized with E. coli-derived Human IFIT3 fragment | |
| 100 μg | |
| Immunology | |
| 3437 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction