missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFIT3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
183.00€ - 322.00€
Specifications
| Antigen | IFIT3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 5-10uL/mg of uLysate |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18639397
|
Novus Biologicals
NBP3-12668-50ul |
50 μg |
183.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18609456
|
Novus Biologicals
NBP3-12668-100ul |
100 μg |
322.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFIT3 Polyclonal antibody specifically detects IFIT3 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| IFIT3 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| PBS, pH7.0 | |
| 3437 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunoprecipitation 5-10uL/mg of uLysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CIG-49, GARG-49, IFI60Retinoic acid-induced gene G protein, IFIT-3, IFIT-4, IFIT4IFI-60K, interferon-induced protein with tetratricopeptide repeats 3, Interferon-induced protein with tetratricopeptide repeats 4Interferon-induced 60 kDa protein, IRG2, ISG60, ISG-60, RIG-GCIG49 | |
| Produced in rabbits immunized with E. coli-derived Human IFIT3 fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title