missing translation for 'onlineSavingsMsg'
Learn More

IFN-gamma Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-11362-0.025mg

 View more versions of this product

Product Code. 18607117

  • 287.00€ / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IFN-gamma Polyclonal antibody specifically detects IFN-gamma in Feline samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IFN-gamma
Polyclonal
PBS
Rabbit
Affinity purified
RUO
Primary
Feline
Purified
Western Blot, ELISA
Unconjugated
IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Recombinant feline IFN Gamma
0.025 mg
Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers
3458
Store at 4°C. Do not freeze.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.