missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-gamma Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
287.00€ - 840.00€
Specifications
| Antigen | IFN-gamma |
|---|---|
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18607117
|
Novus Biologicals
NBP3-11362-0.025mg |
0.025 mg |
287.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610199
|
Novus Biologicals
NBP3-11362-0.1mg |
0.1 mg |
840.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-gamma Polyclonal antibody specifically detects IFN-gamma in Feline samples. It is validated for Western Blot, ELISASpecifications
| IFN-gamma | |
| Polyclonal | |
| Purified | |
| RUO | |
| Feline | |
| IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| Recombinant feline IFN Gamma | |
| Primary | |
| Store at 4°C. Do not freeze. |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers | |
| PBS | |
| 3458 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title