missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFRD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24679
This item is not returnable.
View return policy
Description
IFRD1 Polyclonal specifically detects IFRD1 in Human, Porcine, Bovine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IFRD1 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| IFRD1 | |
| A portion of amino acids 70-120 of human Tis7 was used as the immunogen. | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, cow, horse, dog, chimpanzee, 92% homologous in mouse, rat, macaque, rhesus monkey, and 83% homologous in opossum. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 1.0-4.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| 12-O-tetradecanoylphorbol-13-acetate-induced sequence 7, interferon-related developmental regulator 1, Nerve growth factor-inducible protein PC4, PC4, pheochromocytoma cell-4, TIS7, TPA induced sequence 7 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3475 | |
| Human, Pig, Bovine, Canine, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction