missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFRD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00€ - 361.00€
Specifications
| Antigen | IFRD1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1.0-4.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18126951
|
Novus Biologicals
NBP2-24679SS |
0.025 mg |
213.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18110102
|
Novus Biologicals
NBP2-24679 |
0.1 mg |
361.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFRD1 Polyclonal specifically detects IFRD1 in Human, Porcine, Bovine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IFRD1 | |
| Western Blot 1.0-4.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 3475 | |
| A portion of amino acids 70-120 of human Tis7 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Pig, Bovine, Canine, Primate | |
| 12-O-tetradecanoylphorbol-13-acetate-induced sequence 7, interferon-related developmental regulator 1, Nerve growth factor-inducible protein PC4, PC4, pheochromocytoma cell-4, TIS7, TPA induced sequence 7 | |
| IFRD1 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, cow, horse, dog, chimpanzee, 92% homologous in mouse, rat, macaque, rhesus monkey, and 83% homologous in opossum. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title