missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFT25 Polyclonal Antibody

Description
Antibody detects endogenous levels of total IFT25.
HSPB11 (Heat shock protein beta-11), also known as IFT25, was originally identified as a small heat shock protein. Recently it has been identified as a component of IFT complex B. It has been demonstrated that IFT25 is not required for ciliary assembly but is required for proper Hedgehog signaling, which in mammals occurs within cilia. Cilia lacking IFT25 have defects in the signal-dependent transport of multiple Hedgehog components and fail to activate the pathway upon stimulation.
Specifications
Specifications
| Antigen | IFT25 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Hspb11 |
| Gene Accession No. | Q9D6H2, Q9Y547 |
| Gene Alias | 2900042B11Rik; C1orf41; Heat shock protein; Heat shock protein beta-11; heat shock protein family B (small) member 11; heat shock protein family B (small), member 11; HSP; Hspb11; Hspb11 heat shock protein family B (small), member 11; HSPC034; HSPCO34; IFT25; intraflagellar transport 25 homolog; intraflagellar transport protein 25 homolog; Placental protein 25; placental protein 25 homolog; PP25 |
| Gene Symbols | Hspb11 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?