missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IGFBP-1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16561122
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16561122 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16561122 Supplier Invitrogen™ Supplier No. PA598298

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IGFBP-1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene is a member of the insulin-like growth factor binding protein (IGFBP) family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors (IGFs) I and II and circulates in the plasma. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFBP-1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene IGFBP1
Gene Accession No. P08833
Gene Alias AFBP; alpha-pregnancy-associated endometrial globulin; amniotic fluid binding protein; Binding protein 25; Binding protein 26; Binding protein 28; binding protein-25; binding protein-26; binding protein-28; growth hormone independent-binding protein; hIGFBP 1; hIGFBP1; hIGFBP-1; IBP 1; IBP1; IBP-1; IGF BP25; IGFBA; IGF-binding protein 1; IGFBP; IGFBP 1; Igfbp1; Igfbp-1; IGF-BP25; Insulin like growth factor binding protein; insulin like growth factor binding protein 1; insulin-like growth factor binding protein 1; INSULIN-LIKE GROWTH FACTOR BINDING PROTEIN 1 PRECURSOR (IGFBP-1) (IBP-1) (IGF-BINDING PROTEIN 1); insulin-like growth factor-binding protein 1; Placental protein 12; PP 12; PP12
Gene Symbols IGFBP1
Host Species Rabbit
Immunogen Recombinant Human Insulin-like growth factor-binding protein 1 protein (26-259AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3484
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.