missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgK Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NB100-75170-0.025mg
This item is not returnable.
View return policy
Description
IgK Polyclonal specifically detects IgK in Human samples. It is validated for Western Blot.
Specifications
| IgK | |
| Polyclonal | |
| Western Blot 1:100-1:2000 | |
| IGK@, immunoglobulin kappa locus | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| 50802 | |
| Human |
| Western Blot | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| IGK | |
| Human IgK (Kappa) native protein. | |
| 0.025 mg | |
| Primary | |
| Ig k | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction