missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IHH Polyclonal Antibody
GREENER_CHOICE

Product Code. 30534345
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30534345 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30534345 Supplier Invitrogen™ Supplier No. PA5148524

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Ihh encodes a member of the hedgehog family of secreted signaling molecules. Hedgehog proteins are essential regulators of a variety of developmental processes including growth, patterning and morphogenesis. The encoded protein specifically plays a role in bone growth an differentiation. Mutations in this gene are the cause of brachydactyly type A1 which is characterized by shortening or malformation of the phalanges. Mutations in this gene are also the cause of acrocapitofemoral dysplasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IHH
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene Ihh
Gene Accession No. P97812
Gene Alias BDA1; HHG2; HHG-2; Ihh; Indian hedgehog; Indian hedgehog homolog; indian hedgehog protein; Indian hedgehog protein C-product; Indian hedgehog protein N-product; indian hedgehog protein; LOW QUALITY PROTEIN: indian hedgehog protein; Indian hedgehog signaling molecule; Indian hedgehog-like protein
Gene Symbols Ihh
Host Species Goat
Immunogen E. coli-derived recombinant mouse Indian Hedgehog, Cys28-Gly202.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16147
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.