Learn More
IKZF1 Mouse anti-Human, Unconjugated, Clone: 1D6B5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66966-1-IG-20UL
 View more versions of this product
Product Code. 16809305
  • 145.00€ / 20µL
Description
The IKZF1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IKZF1. This antibody recognizes human, mouse, rabbit, pig antigen. The IKZF1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
IKZF1
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse, Rabbit, Pig
Western Blot
1D6B5
Unconjugated
20 μL
10320