Learn More
IKZF1 Mouse anti-Human, Unconjugated, Clone: 1D6B5, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | IKZF1 |
|---|---|
| Clone | 1D6B5 |
| Concentration | 500 μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16809305
|
Proteintech
66966-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16899295
|
Proteintech
66966-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The IKZF1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IKZF1. This antibody recognizes human, mouse, rabbit, pig antigen. The IKZF1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| IKZF1 | |
| 500 μg/mL | |
| Monoclonal | |
| Mouse | |
| 10320 | |
| Primary |
| 1D6B5 | |
| Western Blot | |
| Unconjugated | |
| Human, Mouse, Rabbit, Pig | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.