Learn More
IL-12 p70 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | IL-12 p70 |
|---|---|
| Concentration | 400 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18037025
|
Proteintech
30756-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18054235
|
Proteintech
30756-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The IL-12 p70 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse IL-12 p70. This antibody recognizes mouse antigen. The IL-12 p70 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| IL-12 p70 | |
| Western Blot, ELISA | |
| Rabbit | |
| 16160 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 400 μg/mL | |
| Liquid | |
| Mouse | |
| IL12, IL-12, IL-12 p70 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.