missing translation for 'onlineSavingsMsg'
Learn More

IL-23R Antibody (753317) [DyLight 405], Novus Biologicals™

Product Code. 18542965 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18542965 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18542965 Supplier Novus Biologicals Supplier No. FAB16861E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-23R Monoclonal antibody specifically detects IL-23R in Mouse samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-23R
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 753317
Conjugate DyLight 405
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias IBD17, IL-23 receptor, IL-23R, interleukin 23 receptor, interleukin-23 receptor
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-23 R, Gly24-Asp372, Accession # Q8K4B4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 149233
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.