missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-6 Antibody (AP-MAB0847) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-06663-0.1mg
This item is not returnable.
View return policy
Description
IL-6 Monoclonal specifically detects IL-6 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications
| IL-6 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready, Block/Neutralize | |
| B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6 | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with Mouse recombinant IL-6. | |
| Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles. | |
| IgG1 |
| Flow Cytometry | |
| AP-MAB0847 | |
| Unconjugated | |
| PBS with No Preservative | |
| IL6 | |
| Mouse recombinant IL-6 | |
| 0.1 mg | |
| Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
| 3569 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction