missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-6 Antibody (AP-MAB0847) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
190.00€
Specifications
| Antigen | IL-6 |
|---|---|
| Clone | AP-MAB0847 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready, Block/Neutralize |
| Applications | Flow Cytometry |
Description
IL-6 Monoclonal specifically detects IL-6 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready.Specifications
| IL-6 | |
| 1 mg/mL | |
| Flow Cytometry | |
| Unconjugated | |
| Rat | |
| Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
| PBS with No Preservative | |
| 3569 | |
| Mouse recombinant IL-6 | |
| Primary | |
| Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles. |
| AP-MAB0847 | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready, Block/Neutralize | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6 | |
| IL6 | |
| IgG1 | |
| Protein A or G purified | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with Mouse recombinant IL-6. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title