missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL1RAPL1 Monoclonal specifically detects IL1RAPL1 in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.
Specifications
Specifications
| Antigen | IL1RAPL1 |
| Applications | Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2H3C12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | IL1R8IL1RAPL, IL-1-RAPL-1, IL-1RAPL-1, IL1RAPL-1, interleukin 1 receptor accessory protein-like 1, interleukin 1 receptor-8, interleukin-1 receptor accessory protein-like 1, mental retardation, X-linked 10, MRX10, MRX21, MRX34, Oligophrenin-4, OPHN4mental retardation, X-linked 21, Three immunoglobulin domain-containing IL-1 receptor-related 2, TIGIRR-2mental retardation, X-linked 34, X-linked interleukin-1 receptor accessory protein-like 1 |
| Gene Symbols | IL1RAPL1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?