Learn More
IL20 Monoclonal Antibody (OTI5E10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803409S
Description
IL20 Monoclonal antibody specifically detects IL20 in Human samples. It is validated for Western BlotSpecifications
| IL20 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H492 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84557 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI5E10 | |
| Unconjugated | |
| MAP1LC3A | |
| cytokine Zcyto10; four alpha helix cytokine; four alpha helix cytokine ZCYTO10; four helix bundle cytokine 10; il 20; IL10D; Il20; IL-20; ILN; Interleukin; interleukin 20; interleukin-20; MGC96907; PRO1801; UNQ852; UNQ852/PRO1801; ZCYTO10 | |
| Full length human recombinant protein of human MAP1LC3A produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.