missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL6R Antibody (B-R6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64770-0.025MG
This item is not returnable.
View return policy
Description
IL6R Monoclonal specifically detects IL6R in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| IL6R | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:50, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| P08887 | |
| IL6R | |
| CHO cells transfected with cDNA of the IL-6 receptor | |
| 0.025 mg | |
| Cancer, Immunology, Innate Immunity | |
| 3570 | |
| Human | |
| Purified |
| Immunofluorescence, Immunohistochemistry, Flow Cytometry, ELISA, Immunoprecipitation, Immunocytochemistry | |
| B-R6 | |
| Unconjugated | |
| PBS (pH 7.4) with 0.09% Sodium Azide | |
| CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes soluble and membranous human IL-6R. This has been shown to inhibit IL-6 mediated proliferation of XG-1 cells, and partially blocks binding of IL-6 to its receptor. Removal of Sodium azide is recommended prior to use in functional assays. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction