missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL6R Antibody (B-R6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
220.00€ - 593.00€
Specifications
| Antigen | IL6R |
|---|---|
| Clone | B-R6 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:50, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Immunofluorescence, Immunohistochemistry, Flow Cytometry, ELISA, Immunoprecipitation, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18027753
|
Novus Biologicals
NB100-64770-0.025MG |
0.025 mg |
220.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18060916
|
Novus Biologicals
NB100-64770 |
0.1 mg |
593.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL6R Monoclonal specifically detects IL6R in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| IL6R | |
| 1 mg/mL | |
| Immunofluorescence, Immunohistochemistry, Flow Cytometry, ELISA, Immunoprecipitation, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, Immunology, Innate Immunity | |
| P08887 | |
| 3570 | |
| CHO cells transfected with cDNA of the IL-6 receptor | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| B-R6 | |
| Flow Cytometry 1:50, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991 | |
| IL6R | |
| IgG1 | |
| Protein A or G purified | |
| Recognizes soluble and membranous human IL-6R. This has been shown to inhibit IL-6 mediated proliferation of XG-1 cells, and partially blocks binding of IL-6 to its receptor. Removal of Sodium azide is recommended prior to use in functional assays. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title