missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IMP4 Polyclonal Antibody

Product Code. 15909544
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15909544 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15909544 Supplier Invitrogen™ Supplier No. PA566858

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: NRLIPTELRRE ALALQGSLEF DDAGGEGVTS HVDDEYRWAG VEDPKVMITT SRDPSSRLKM FAKE Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.

IMP4 forms a ternary complex with IMP3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IMP4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Imp4
Gene Accession No. Q96G21
Gene Alias AA409888; AV031295; Brix domain-containing protein 4; BXDC4; D1Wsu40e; IMP4; IMP4 homolog, U3 small nucleolar ribonucleoprotein; IMP4, U3 small nucleolar ribonucleoprotein; IMP4, U3 small nucleolar ribonucleoprotein, homolog; IMP4, U3 small nucleolar ribonucleoprotein, homolog (yeast); LRRGT00017; RGD1308463; U3 small nucleolar ribonucleoprotein protein IMP4; U3 snoRNP protein 4 homolog; U3 snoRNP protein IMP4
Gene Symbols Imp4
Host Species Rabbit
Immunogen Recombinant Human IMP4. Recombinant protein control fragment (Product #RP-107511).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92856
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.